Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrodes
pH Electrodes
Accessories for Lovibond® Water Testing pH Pocket Tester, SD10
Accessories for Lovibond® Water Testing pH Pocket Tester, SD10
Accessories for Lovibond® Water Testing pH Pocket Tester, SD10
Catalog # LOVI720015
Supplier:
Lovibond
Expand image
Accessories for Lovibond® Water Testing pH Pocket Tester, SD10
Catalog # LOVI720015
Supplier:
Lovibond
Specifications
Description:
Replacement electrode for SD10 pH
Pk:
1
Specifications
About this item
Product Details
Product Details