

Acetic acid
Catalog # VWRS5257.9190
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic drum
- Pk:210 L
- Seq:5585
Specifications
About this item
:
:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
:GHS02
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152