Tetrabromophthalic anhydride, Sigma-Aldrich®
Catalog # SIALT5206-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:463.7 g/mol
Boiling Pt:477 °C (1013 hPa)
Melting Pt:271…274 °C
Density:2,93 g/cm³ (20 °C)
EINECS:211-185-4
CAS Number:632-79-1
Formula:C₈Br₄O₃
MDL Number:MFCD00005919