3,3',5,5'-Tetramethylbenzidine (TMB) ≥98% (by TLC), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:54827-17-7
:
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748
:240.35 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:166…170 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:259-364-6
Frequently Bought Together