3-Brompropylamine hydrobromide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5003-71-4
:
Formula:BrH₂CCH₂CH₂NH₃Br
MDL Number:MFCD00012912
:218.92 g/mol
Melting Pt:173…174 °C
:M-Clarity™ quality level = MQ200
EINECS:225-675-0
:GHS07
Frequently Bought Together