Pig Pepsin (from Gastric mucosa)
Catalog # SIAL77160-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- CAS No.:9001-75-6
- Source:Gastric mucosa
- Species:Pig
- Storage conditions:Refrigerator
- Enzyme name:Pepsin
- Enzyme activity:50 U/g