1-Hexyl-3-methylimidazolium tetrafluoroborate, Sigma-Aldrich®
SIAL73244-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:254.08 g/mol
:∼200 °C (1013 hPa)
:–82 °C
:1.14 g/cm³ (20 °C)
:>100 °C
:244193-50-8
:C₁₀H₁₉BF₄N₂
:MFCD03095459