Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tetramethyl(propyl)cyclopentadiene
Tetramethyl(propyl)cyclopentadiene, Sigma-Aldrich®
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
Tetramethyl(n-propyl)cyclopentadiene
:
:
164.29 g/mol
:
3,III
:
M-Clarity™ quality level = MQ100
:
3295
:
64417-12-5
:
C₁₂H₂₀
:
MFCD00145386