

(Methoxymethyl)triphenylphosphonium chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4009-98-7
:
Formula:C₂₀H₂₀ClOP
MDL Number:MFCD00011800
:342.8 g/mol
Melting Pt:190 °C
:M-Clarity™ quality level = MQ100
EINECS:223-664-5
:GHS07
:GHS09
Frequently Bought Together