2-(4-Methoxyphenyl)ethanol, Sigma-Aldrich®
Catalog # SIAL154180-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:152.19 g/mol
:M-Clarity™ quality level = MQ100
EINECS:211-866-6
CAS Number:702-23-8
Formula:C₉H₁₂O₂
MDL Number:MFCD00002900