N-Isopropylbenzylamine, Sigma-Aldrich®
Catalog # SIAL136964-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:149.24 g/mol
Boiling Pt:200 °C (1013 hPa)
Melting Pt:∼ 0 °C
Density:0.89 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:203-067-6
UN:2735
ADR:8,III
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:102-97-6
Formula:C₆H₅CH₂N(CH₃)(CH₂CH₃)
MDL Number:MFCD00008863