6-Methoxy-m-toluidine, Sigma-Aldrich®
Catalog # SIAL103284-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-Amino-4-methoxytoluene, 2-Amino-4-methylanisole, 1-Amino-2-methoxy-5-methylbenzene, 5-Methyl-o-anisidine, p-Cresidine, 2-Methoxy-5-methylaniline
:
:137.18 g/mol
Boiling Pt:235 °C (1013 hPa)
Melting Pt:55.5 °C
Density:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:204-419-1
:GHS08
:GHS07
CAS Number:120-71-8
Formula:C₈H₁₁NO
MDL Number:MFCD00007815