Order Entry
Slovenia
ContactUsLinkComponent
2-Chlorobenzoxazole
2-Chlorobenzoxazole
Catalog # MOLEM18514150
Supplier:  Molekula
CAS Number:  
2-Chlorobenzoxazole
Catalog # MOLEM18514150
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Boiling Pt:202 °C (1013 hPa)
Melting Pt:45 °C
Density:1.3453 g/cm³ (18 °C)
 :153.57 g/mol
EINECS:210-414-5
CAS Number:615-18-9
Formula:C₇H₄ClNO
MDL Number:MFCD00005766
Boiling Pt:202 °C (1013 hPa)