Order Entry
Slovenia
ContactUsLinkComponent
Disodium succinate for synthesis, Sigma-Aldrich®
Disodium succinate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Disodium succinate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    350 g/l (25 °C)
  • Melting Point:
    300 °C
  • Molar Mass:
    162.05 g/mol
  • pH:
    7 - 9 (100 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together