

DL-Camphor
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:76-22-2
:
Formula:C₁₀H₁₆O
MDL Number:MFCD00074738
:152.24 g/mol
Boiling Pt:204 °C (1013 hPa)
Melting Pt:∼180 °C
Density:0.992 g/cm³ (25 °C)
Flash Pt:66 °C
Storage Temperature:Ambient
EINECS:200-945-0
UN:2717
ADR:4.1,III
:GHS02
:GHS07
Frequently Bought Together