2-Fluorocyclohexanone 96%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 96% CAS No.: 694-82-6 Molecular Formula: C6H9FO Formula Weight: 116.13 Storage Temperature: -30°C to -10°C Flash Point: 71°(160°F) Refractive Index: 1.4440 Boiling Point: 78°/17mm MDL No.: MFCD01076476
- CAS Number:694-82-6
- Formula:C₆H₉FO
- MDL Number:MFCD01076476
- Molecular Weight:116.14 g/mol
- Boiling Pt:78 °C (17 mmHg)
- Density:1.08
- Flash Pt:71 °C (160 °F)
2 Options Available Below
Product Details & Documents
Purity: 96% CAS No.: 694-82-6 Molecular Formula: C6H9FO Formula Weight: 116.13 Storage Temperature: -30°C to -10°C Flash Point: 71°(160°F) Refractive Index: 1.4440 Boiling Point: 78°/17mm MDL No.: MFCD01076476
- CAS Number:694-82-6
- Formula:C₆H₉FO
- MDL Number:MFCD01076476
- Molecular Weight:116.14 g/mol
- Boiling Pt:78 °C (17 mmHg)
- Density:1.08
- Flash Pt:71 °C (160 °F)



