Order Entry
Slovenia
ContactUsLinkComponent
2-(4-Methoxyphenyl)ethanol 96%
2-(4-Methoxyphenyl)ethanol 96%
Certificates
undefined
2-(4-Methoxyphenyl)ethanol 96%

Specification Test Results

  • Appearance:
    Clear colorless to light brown Liquid or solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5360 to 1.5380 (20°C, 589 nm)
  • GC:
    ≥95 %

Specification Test Results

Related Information

Frequently Bought Together