Ethyl chloroacetate 99%
:105-39-5
:
:C₄H₇ClO₂
:MFCD00000932
:13,03818
:142…145 °C (760 mmHg)
:–26 °C
:1.15 g/cm³
:53 °C
:Ambient
:1181
:6.1,II
:122.55 g/mol
:203-294-0
:GHS06
:GHS09
:GHS02
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4200 to 1.4220 (20°C, 589 nm)
- GC:≥98.5 %
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.005 %
Frequently Bought Together