Protease (from Bacillus licheniformis)
Catalog # ANKOTDF83
Supplier: ANKOM TECHNOLOGY
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:100 mL
- CAS No.:79986-26-8
- Storage conditions:Store at 4 °C
- Enzyme name:Protease (from Bacillus licheniformis)
- Enzyme synonyms:Subtilisin A
Specifications
About this item
This enzyme formulation was developed specifically for use in the ANKOMTDF dietary fiber analyser. With all of the necessary ingredients in this concentrated formula, just add the correct amount of water and the enzyme solution is ready for use in the instrument. This product has enough enzyme for approximately 500 ANKOMTDF dietary fiber analyser assays.
Contents: 100 ml; 175 Tyrosine U/ml; in 50% glycerol + 0,09% sodium azide. From Bacillus licheniformis. Highly purified.