0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Tide Fluor™ 6WS alkyne
Tide Fluor™ 6WS alkyne, Tide Fluor™ fluorescent probe
Tide Fluor™ 6WS alkyne, Tide Fluor™ fluorescent probe
AATB2303
:
AAT Bioquest
:
Tide Fluor™ 6WS alkyne, Tide Fluor™ fluorescent probe
AATB2303
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
1162.23 g/mol
:
5,II
:
GHS07