Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(Iodoacetamido)fluorescein
5-(Iodoacetamido)fluorescein ≥75%
5-(Iodoacetamido)fluorescein ≥75%
AATB222
:
AAT BIOQUEST
:
5-(Iodoacetamido)fluorescein ≥75%
AATB222
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
25 mg
Seq:
2315
:
:
515.25 g/mol
:
5,II
:
GHS07
:
63368-54-7
:
C₂₂H₁₄INO₆