Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Violet 510 acid
Violet 510 acid, mFluor™ fluorescent probe
Violet 510 acid, mFluor™ fluorescent probe
Irritant
Violet 510 acid, mFluor™ fluorescent probe
Catalog # AATB1141
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
Violet 510 acid, mFluor™ fluorescent probe
Catalog # AATB1141
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
500 kDa
ADR:
5,II
:
GHS07
Product Details