Native Streptavidin (from Streptomyces avidinii), Alexa Fluor® 555
Supplier: AAT BIOQUEST
Certificates
About this item
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.
- Bright signal
- More photostable fluorescence
- Improved proprietary fluorescent label
Caution: For research use only.
Specifications
- Conjugation:Alexa Fluor® 555
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Protein synonyms:Avidin| Streptomyces avidinii
Specifications
Frequently Bought Together