Order Entry
Slovenia
ContactUsLinkComponent
 
2,3,7,8-Tetrachlorodibenzo-p-dioxin
Catalog # CAYM38392-1
Supplier:  Cayman Chemical
CAS Number:  
2,3,7,8-Tetrachlorodibenzo-p-dioxin
Catalog # CAYM38392-1
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    1 mg
  • CAS No.:
    1746-01-6
  • Description:
    2,3,7,8-Tetrachlorodibenzo-p-dioxin
  • Hazard class description:
    6.1 Toxic substances
  • Shipping conditions:
    Room temperature in continental US; may vary elsewhere
  • UN No.:
    2811
  • GHS signal:
    Danger
  • Storage temperature:
    −20 °C

Specifications

About this item