Order Entry
Slovenia
ContactUsLinkComponent
Phalloidin, XFD350
Phalloidin, XFD350
Catalog # AATB23150
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Phalloidin, XFD350
Catalog # AATB23150
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    300 Tests
  • Conjugation:
    Alexa Fluor® 350
  • Protein/peptide type:
    Native
  • Source:
    Amanita phalloides
  • Species:
    Amanita phalloides
  • Protein synonyms:
    heptapeptide phallotoxin
  • Protein/peptide name:
    Phalloidin

Specifications

About this item

XFD350 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 350 (Alexa Fluor® is the trademark of ThermoFisher).

  • Well-cited conjugate
  • High affinity and selectivity
  • In house quality control
Caution: For research use only.