Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), IRDye® 800
Catalog # AATB16977
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Vial
- Pk:200 µG
- Conjugation:IRDye® 800
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Species:Streptomyces avidinii
- Protein synonyms:Avidin|Streptomyces avidinii
- Protein/peptide name:Streptavidin
Specifications
About this item
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.
- Extremely high biotin-binding affinity
- High quality conjugate
Caution: For research use only.