Order Entry
Slovenia
ContactUsLinkComponent
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 594
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 594
Supplier:  AAT BIOQUEST
Certificates
undefined
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 594
Supplier:  AAT BIOQUEST

Specifications

  • Conjugation:
    iFluor® 594
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Species:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin|Streptomyces avidinii
  • Protein/peptide name:
    Streptavidin

Specifications

Frequently Bought Together