Order Entry
Slovenia
ContactUsLinkComponent
HRP-Labeled Goat Anti-Mouse IgG (H+L) (1 mg)
HRP-Labeled Goat Anti-Mouse IgG (H+L) (1 mg)
  AATB16728
undefined
HRP-Labeled Goat Anti-Mouse IgG (H+L) (1 mg)
  AATB16728

 

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Clonality:
    Polyclonal
  • Clone:
    IgG
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Goat
  • Isotype:
    IgG (H, L)
  • Reactivity:
    Mouse
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

The enzyme labels can be visualized by means of enzyme histochemical methods via chromogenic (producing colour) reactions, or via fluorogenic (producing fluorescence) reactions.

  • Very widely used
  • High activity
  • Compatible with all major platforms
  • Multiple applications
 : For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse