This gene encodes a subunit of the asialoglycoprotein receptor. This receptor is a transmembrane protein that plays a critical role in serum glycoprotein homeostasis by mediating the endocytosis and lysosomal degradation of glycoproteins with exposed terminal galactose or N-acetylgalactosamine residues.
- Expression host: HEK293T
- Buffer: 25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol
Stable for 12 months from the date of receipt of the product under proper storage and handling conditions. Avoid repeated freeze-thaw cycles.
The asialoglycoprotein receptor may facilitate hepatic infection by multiple viruses including hepatitis B, and is also a target for liver-specific drug delivery. The asialoglycoprotein receptor is a hetero-oligomeric protein composed of major and minor subunits, which are encoded by different genes. The protein encoded by this gene is the more abundant major subunit. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
- Protein/peptide type:Recombinant
- Species:Human
- Tag sequence:C-Myc/DDK
- Storage conditions:Store at −80 °C
- Protein synonyms:ASGPR|ASGPR1|HL-1|CLEC4H1|Asialoglycoprotein Receptor 1
- UniProtKB:P17661, P07306, Q53SB5, Q6FGQ5
- Protein/peptide name:ASGR1
- Purity:>80% as determined by SDS-PAGE and Coomassie blue staining
- Molecular weight:33 kDa
- Sequence:MTKEYQDLQHLDNEESDHHQLRKGPPPPQPLLQRLCSGPRLLLLSLGLSLLLLVVVCVIGSQNSQLQEEL
RGLRETFSNFTASTEAQVKGLSTQGGNVGRKMKSLESQLEKQQKDLSEDHSSLLLHVKQFVSDLRSLSCQ
MAALQGNGSERTCCPVNWVEHERSCYWFSRSGKAWADADNYCRLEDAHLVVVTSWEEQKFVQHHIGPVNT
WMGLHDQNGPWKWVDGTDYETGFKNWRPEQPDDWYGHGLGGGEDCAHFTDDGRWNDDVCQRPYRWVCETE
LDKASQEPPLL
TRTRPLEQKLISEEDLAANDILDYKDDDDKV - Concentration:>0,05 µg/µl as determined by microplate BCA method