Order Entry
Slovenia
ContactUsLinkComponent
Tide Fluor™ 3WS acid, Tide Fluor™ fluorescent probe
Irritant
Tide Fluor™ 3WS acid, Tide Fluor™ fluorescent probe
Catalog # AATB2345
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Tide Fluor™ 3WS acid, Tide Fluor™ fluorescent probe
Catalog # AATB2345
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :838.89 g/mol
ADR:5,II
 :GHS07