Order Entry
Slovenia
ContactUsLinkComponent
TRITC-DHPE ≥90%
TRITC-DHPE ≥90%
Catalog # AATB23301
Supplier:  AAT Bioquest
CAS Number:  
undefined
TRITC-DHPE ≥90%
Catalog # AATB23301
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1236.67 g/mol
CAS Number:176181-90-1
Formula:C₆₈H₁₁₀N₅O₁₁PS