Order Entry
Slovenia
ContactUsLinkComponent
 
Irritant
Rhodamine 123 ≥95%
Catalog # AATB22210
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Rhodamine 123 ≥95%
Catalog # AATB22210
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
ADR:5,II
 :380.82 g/mol
 :GHS07
CAS Number:62669-70-9
Formula:C₂₁H₁₇ClN₂O₃
MDL Number:MFCD00012664