Order Entry
Slovenia
ContactUsLinkComponent
3,3-Diheptyloxacarbocyanine iodide [DiOC7(3) iodide] ≥95%
3,3-Diheptyloxacarbocyanine iodide [DiOC7(3) iodide] ≥95%
Catalog # AATB22040
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
3,3-Diheptyloxacarbocyanine iodide [DiOC7(3) iodide] ≥95%
Catalog # AATB22040
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :600.57 g/mol