Order Entry
Slovenia
ContactUsLinkComponent
Sodium bromate ≥98%, TECHNICAL
Oxidizing
Irritant
Sodium bromate ≥98%, TECHNICAL
Catalog # 27734.460
Supplier:  VWR Chemicals
CAS Number:  
undefined
Oxidizing
Irritant
Sodium bromate ≥98%, TECHNICAL
Catalog # 27734.460
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bucket
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

About this item

 :
 :150.89 g/mol
Merck Index:12,08736
Melting Pt:381 °C
Storage Temperature:Ambient
EINECS:232-160-4
UN:1494
ADR:5.1,II
 :GHS03
 :GHS07
CAS Number:7789-38-0
Formula:BrNaO₃
MDL Number:MFCD00003476