Order Entry
Slovenia
ContactUsLinkComponent
Nordic ID Stix UHF RFID Reader + Writer (USB) EU
Nordic ID Stix UHF RFID Reader + Writer (USB) EU
Catalog # BRDY311095
Supplier:  Brady
undefined
Nordic ID Stix UHF RFID Reader + Writer (USB) EU
Catalog # BRDY311095
Supplier:  Brady
Specifications
  • Description:
    Stix UHF RFID (USB) EU
  • W×D×H:
    53×79×7 mm
  • Interfaces:
    USB 2.0
  • Pk:
    1
Specifications
About this item
Extra small UHF RFID reader + writer with USB connectivity.
  • Suitable for Android devices
  • Easy installation
  • USB powered
  • No battery
This Stix is an affordable credit card sized UHF RFID reader and writer with USB connectivity. It allows an easy way of bringing RFID capabilities into your PC or laptop.