0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Proteins and Peptides
Recombinant Human VEGFA (from HEK293 cells)
Recombinant Human VEGFA (from HEK293 cells)
ABNOP9303.10UG
:
Abnova
:
Recombinant Human VEGFA (from HEK293 cells)
ABNOP9303.10UG
:
Abnova
:
Pk:
10 µG
Protein/peptide type:
Recombinant
Source:
HEK293 cells
Species:
Human
Protein/peptide name:
VEGFA