Recombinant Human POSTN (from HEK293 cells), His-tag
Catalog # ABNOP9025.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His-tag
- Gene ID:Q15063
- Protein/peptide name:POSTN
- Sequence:22a.a.-836a.a.