Recombinant Human SPRED1 (from E. coli), His-tag
Catalog # ABNOP7704.250UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:250 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:NP_689807
- Protein/peptide name:SPRED1
- Sequence:1a.a.-444a.a