Recombinant Human RBM18 (from E. coli), His-tag
Catalog # ABNOP7707.500UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:500 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Gene ID:NP_149108
- Protein/peptide name:RBM18
- Sequence:1a.a.-190a.a