0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Proteins and Peptides
Recombinant Mouse VEGFA
Recombinant Mouse VEGFA
Catalog # ABNOP9315.100UG
Supplier:
Abnova
CAS Number:
Expand image
Recombinant Mouse VEGFA
Catalog # ABNOP9315.100UG
Supplier:
Abnova
CAS Number:
Specifications
Pk:
100 µG
Protein/peptide type:
Recombinant
Species:
Mouse
Protein/peptide name:
VEGFA
Specifications
About this item
Product Details
Product Details