Recombinant Human PDGFD (from CHO cells)
Catalog # ABNOP7434.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Gene ID:Q9GZP0
- Protein/peptide name:PDGFD
- Sequence:250a.a.-370a.a.