Order Entry
Slovenia
ContactUsLinkComponent
Versilic™ Tubings, Silicone
undefined
Versilic™ Tubings, Silicone
  • Catalog No:
  • VERN760670
  • 228-0376
  • 228-0379
  • VERN760510
  • VERN760500
  • VERN760520
  • 228-0373
  • Max. temperature:
  • 200 °C
  • 200 °C
  • 200 °C
  • 200 °C
  • 200 °C
  • 200 °C
  • 200 °C
  • Min. temperature:
  • −50 °C
  • −50 °C
  • −50 °C
  • −50 °C
  • −50 °C
  • −50 °C
  • −50 °C
Frequently Bought Together