Anti-StAR Rabbit Recombinant Antibody [clone: R04-5G9]
Catalog # ABNORAB01695.100UL
Supplier: Abnova
Specifications
- Antigen symbol:StAR
- Clonality:Recombinant
- Clone:R04-5G9
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human StAR
- Pk:100 µl