Order Entry
Slovenia
ContactUsLinkComponent
Anti-HDAC2 Rabbit Recombinant Antibody [clone: R06-4E4]
Anti-HDAC2 Rabbit Recombinant Antibody [clone: R06-4E4]
Catalog # ABNORAB02231.100UL
Supplier:  Abnova
Anti-HDAC2 Rabbit Recombinant Antibody [clone: R06-4E4]
Catalog # ABNORAB02231.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    HDAC2
  • Clonality:
    Recombinant
  • Clone:
    R06-4E4
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human HDAC2.
  • Pk:
    100 µl

Specifications

About this item