Order Entry
Slovenia
ContactUsLinkComponent
Anti-GSTP1 Rabbit Recombinant Antibody [clone: R09-4G8]
Anti-GSTP1 Rabbit Recombinant Antibody [clone: R09-4G8]
Catalog # ABNORAB02235.100UL
Supplier:  Abnova
Anti-GSTP1 Rabbit Recombinant Antibody [clone: R09-4G8]
Catalog # ABNORAB02235.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    GSTP1
  • Clonality:
    Recombinant
  • Clone:
    R09-4G8
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human GSTP1.
  • Pk:
    100 µl

Specifications

About this item