Anti-GNAQ Rabbit Recombinant Antibody [clone: R05-5D7]
Catalog # ABNORAB02243.100UL
Supplier: Abnova
Specifications
- Antigen symbol:GNAQ
- Clonality:Recombinant
- Clone:R05-5D7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human GNAQ.
- Pk:100 µl