Anti-GSTM1 Rabbit Recombinant Antibody [clone: R03-8G2]
ABNORAB02247.100UL
: Abnova
- Antigen symbol:GSTM1
- Clonality:Recombinant
- Clone:R03-8G2
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human GSTM1.
- Pk:100 µl