Order Entry
Slovenia
ContactUsLinkComponent
Anti-H3-3A Rabbit Recombinant Antibody [clone: R04-5J3]
Anti-H3-3A Rabbit Recombinant Antibody [clone: R04-5J3]
Catalog # ABNORAB02220.100UL
Supplier:  Abnova
Anti-H3-3A Rabbit Recombinant Antibody [clone: R04-5J3]
Catalog # ABNORAB02220.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    H3-3A
  • Clonality:
    Recombinant
  • Clone:
    R04-5J3
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human H3-3A.
  • Pk:
    100 µl

Specifications

About this item