Order Entry
Slovenia
ContactUsLinkComponent
Anti-GCLC Rabbit Recombinant Antibody [clone: R03-9D9]
Anti-GCLC Rabbit Recombinant Antibody [clone: R03-9D9]
  ABNORAB02263.100UL
 :  Abnova
Anti-GCLC Rabbit Recombinant Antibody [clone: R03-9D9]
  ABNORAB02263.100UL
 :  Abnova

 

  • Antigen symbol:
    GCLC
  • Clonality:
    Recombinant
  • Clone:
    R03-9D9
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human GCLC.
  • Pk:
    100 µl