Anti-GAP43 Rabbit Recombinant Antibody [clone: R01-4F6]
Catalog # ABNORAB02265.100UL
Supplier: Abnova
Specifications
- Antigen symbol:GAP43
- Clonality:Recombinant
- Clone:R01-4F6
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human GAP43.
- Pk:100 µl